siRNA Details
| Gene Symbol: LOC101928002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110072.1 | 4 | 840 | |||
| GenBank mRNA | BC042465.1 | - | 4 | 840 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501387.5 | - | 4 | 840 | Chr. 12 - 69713633 - 69738568 [-] on Build GRCh38 | |

LOC101928002
Genomic Map