siRNA Details
Gene Symbol: LOC439933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122079.1 | 5 | 757 | |||
NR_122080.1 | 3 | 632 | ||||
GenBank mRNA | BC030111.1 | - | 3 | 638 | ||
BC037904.2 | - | 5 | 757 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC439933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122079.1 | 5 | 757 | |||
NR_122080.1 | 3 | 632 | ||||
GenBank mRNA | BC030111.1 | - | 3 | 638 | ||
BC037904.2 | - | 5 | 757 |