siRNA Details
Gene Symbol: LOC102546294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105057.1 | 4 | 1024 | |||
GenBank mRNA | AK054753.1 | - | 4 | 957 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501695.3 | - | 4 | 957 | Chr. 5 - 148268307 - 148383783 [-] on Build GRCh38 |
