siRNA Details
| Gene Symbol: CARS-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093033.1 | - | 1 | 1687 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499962.1 | - | 1 | 1687 | Chr. 11 - 3029394 - 3041260 [+] on Build GRCh38 | |

CARS-AS1
Genomic Map