siRNA Details
| Gene Symbol: MIR4458HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039989.1 | 2 | 2843 | |||
| GenBank mRNA | BX648541.1 | - | 2 | 2839 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000606459.1 | - | 2 | 2839 | Chr. 5 - 8457691 - 8463095 [+] on Build GRCh38 | |

MIR4458HG
Genomic Map