siRNA Details
Gene Symbol: STX18-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037888.1 | 1 | 261 | |||
GenBank mRNA | BC025734.1 | - | 1 | 188 | ||
DA139592.1 | - | 1 | 261 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: STX18-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037888.1 | 1 | 261 | |||
GenBank mRNA | BC025734.1 | - | 1 | 188 | ||
DA139592.1 | - | 1 | 261 |