siRNA Details
Gene Symbol: HHIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720277.3 | XP_006720340.1 | 2 | 329 | ||
XM_011537236.2 | XP_011535538.1 | 2 | 329 | |||
XM_017021708.1 | XP_016877197.1 | 2 | 329 | |||
GenBank mRNA | BC039687.1 | - | 2 | 249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000502101.2 | - | 2 | 249 | Chr. 14 - 99604556 - 99625740 [+] on Build GRCh38 |
