siRNA Details
Gene Symbol: LOC100128386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133571.1 | 2 | 1277 | |||
GenBank mRNA | AK124326.1 | - | 2 | 1277 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000501397.1 | - | 2 | 1277 | Chr. 11 - 100684162 - 100687955 [-] on Build GRCh38 |
