siRNA Details
| Gene Symbol: LOC105377621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136202.1 | 5 | 642 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505709.1 | - | 2 | 484 | Chr. 4 - 102418602 - 102431282 [-] on Build GRCh38 | |

LOC105377621
Genomic Map