siRNA Details
Gene Symbol: LOC101928509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125896.1 | 2 | 185 | |||
GenBank mRNA | AK125257.1 | - | 2 | 185 | ||
DA617431.1 | - | 2 | 199 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125896.1 | 2 | 185 | |||
GenBank mRNA | AK125257.1 | - | 2 | 185 | ||
DA617431.1 | - | 2 | 199 |