siRNA Details
Gene Symbol: LOC101928509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125896.1 | 5 | 855 | |||
GenBank mRNA | AK125257.1 | - | 5 | 855 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000515444.5 | - | 5 | 855 | Chr. 4 - 174094660 - 174220398 [-] on Build GRCh38 |
