siRNA Details
Gene Symbol: LOC101927849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125887.1 | 7 | 1574 | |||
GenBank mRNA | BC040325.1 | - | 7 | 1574 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000503100.1 | - | 7 | 1574 | Chr. 4 - 149154295 - 149278123 [+] on Build GRCh38 |
