siRNA Details
| Gene Symbol: TRIM52-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102762.1 | 3 | 650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM52-AS1| Gene Symbol: TRIM52-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102762.1 | 3 | 650 | |||