siRNA Details
| Gene Symbol: TRIM52-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102759.1 | 2 | 556 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000514146.1 | - | 2 | 556 | Chr. 5 - 181261212 - 181264261 [+] on Build GRCh38 | |

TRIM52-AS1
Genomic Map