siRNA Details
| Gene Symbol: LOC101927543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125419.1 | 4 | 1472 | |||
| GenBank mRNA | AK057448.1 | - | 4 | 1472 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000509350.1 | - | 4 | 1472 | Chr. 8 - 120761253 - 120776832 [+] on Build GRCh38 | |

LOC101927543
Genomic Map