siRNA Details
| Gene Symbol: TUB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120537.1 | 4 | 1903 | |||
| NR_120538.1 | 3 | 1846 | ||||
| GenBank mRNA | AK055772.1 | - | 4 | 1902 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000506601.1 | - | 4 | 1903 | Chr. 11 - 8060038 - 8069373 [+] on Build GRCh38 | |

TUB-AS1
Genomic Map