siRNA Details
| Gene Symbol: TUB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120537.1 | 3 | 218 | |||
| GenBank mRNA | AK055772.1 | - | 3 | 218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUB-AS1| Gene Symbol: TUB-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120537.1 | 3 | 218 | |||
| GenBank mRNA | AK055772.1 | - | 3 | 218 | ||