siRNA Details
| Gene Symbol: LINC00504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126435.1 | 6 | 525 | |||
| GenBank mRNA | BC036758.2 | - | 6 | 536 | 5563071 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00504| Gene Symbol: LINC00504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126435.1 | 6 | 525 | |||
| GenBank mRNA | BC036758.2 | - | 6 | 536 | 5563071 | |