siRNA Details
| Gene Symbol: LINC01197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034095.1 | 4 | 803 | |||
| GenBank mRNA | BC040875.1 | - | 4 | 803 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000508732.6 | - | 4 | 803 | Chr. 15 - 95279284 - 95327100 [-] on Build GRCh38 | |

LINC01197
Genomic Map