siRNA Details
| Gene Symbol: LOC101929544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134273.1 | 1 | 397 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505844.1 | - | 1 | 397 | Chr. 5 - 17436737 - 17441678 [+] on Build GRCh38 | |

LOC101929544
Genomic Map