siRNA Details
| Gene Symbol: LOC101929745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134272.1 | 2 | 1561 | |||
| GenBank mRNA | BC028978.1 | - | 2 | 1561 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000508853.1 | - | 2 | 1561 | Chr. 5 - 38148480 - 38153715 [+] on Build GRCh38 | |

LOC101929745
Genomic Map