siRNA Details
Gene Symbol: LOC101928651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109910.1 | 8 | 1517 | |||
GenBank mRNA | BC043261.1 | - | 8 | 1522 | ||
BC043516.1 | - | 3 | 373 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109910.1 | 8 | 1517 | |||
GenBank mRNA | BC043261.1 | - | 8 | 1522 | ||
BC043516.1 | - | 3 | 373 |