siRNA Details
| Gene Symbol: LINC01339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120601.1 | 6 | 522 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505712.5 | - | 6 | 389 | Chr. 5 - 90158339 - 90289938 [-] on Build GRCh38 | |

LINC01339
Genomic Map