siRNA Details
Gene Symbol: LINC01339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120601.1 | 4 | 304 | |||
GenBank mRNA | BX105792.1 | - | 4 | 279 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505712.5 | - | 4 | 171 | Chr. 5 - 90158339 - 90289938 [-] on Build GRCh38 |
