siRNA Details
Gene Symbol: LOC101929237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125433.1 | 3 | 1564 | |||
GenBank mRNA | AK124682.1 | - | 3 | 1552 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000511897.5 | - | 3 | 1552 | Chr. 8 - 22877984 - 22888022 [+] on Build GRCh38 |
