siRNA Details
| Gene Symbol: LINC01207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038834.1 | 3 | 1972 | |||
| GenBank mRNA | AK095255.1 | - | 3 | 1836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01207| Gene Symbol: LINC01207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038834.1 | 3 | 1972 | |||
| GenBank mRNA | AK095255.1 | - | 3 | 1836 | ||