siRNA Details
| Gene Symbol: LOC101928443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120529.1 | 1 | 635 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000511013.2 | - | 1 | 635 | Chr. 11 - 70056230 - 70065371 [-] on Build GRCh38 | |

LOC101928443
Genomic Map