siRNA Details
Gene Symbol: LINC01574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108030.1 | 2 | 406 | |||
GenBank mRNA | CK906254.1 | - | 1 | 381 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000512115.2 | - | 2 | 406 | Chr. 5 - 176743205 - 176743871 [+] on Build GRCh38 |
