siRNA Details
| Gene Symbol: LINC01574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108030.1 | 2 | 548 | |||
| GenBank mRNA | CK906254.1 | - | 1 | 523 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000512115.2 | - | 2 | 548 | Chr. 5 - 176743205 - 176743871 [+] on Build GRCh38 | |

LINC01574
Genomic Map