siRNA Details
| Gene Symbol: LOC340107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037896.1 | 3 | 1324 | |||
| GenBank mRNA | BC040315.1 | - | 3 | 1324 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000504339.1 | - | 3 | 1324 | Chr. 5 - 24835280 - 24840583 [-] on Build GRCh38 | |

LOC340107
Genomic Map