siRNA Details
| Gene Symbol: LOC100507291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121608.1 | 3, 4 | 358 | |||
| GenBank mRNA | BX098847.1 | - | 3, 4 | 352 | ||
| DB034152.1 | - | 3, 4 | 358 | |||
| DB446595.1 | - | 3, 4 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507291| Gene Symbol: LOC100507291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121608.1 | 3, 4 | 358 | |||
| GenBank mRNA | BX098847.1 | - | 3, 4 | 352 | ||
| DB034152.1 | - | 3, 4 | 358 | |||
| DB446595.1 | - | 3, 4 | 359 | |||