siRNA Details
Gene Symbol: LOC101929470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125817.1 | 3 | 866 | |||
GenBank mRNA | AK054638.1 | - | 3 | 866 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000507496.5 | - | 3 | 866 | Chr. 8 - 29815004 - 29828504 [-] on Build GRCh38 |
