siRNA Details
| Gene Symbol: CTB-113P19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109873.1 | 5 | 1645 | |||
| GenBank mRNA | BC039364.1 | - | 5 | 1646 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000510576.6 | - | 5 | 1645 | Chr. 5 - 151676945 - 151687910 [+] on Build GRCh38 | |

CTB-113P19.1
Genomic Map