siRNA Details
| Gene Symbol: LOC101929380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105018.1 | 2 | 187 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929380| Gene Symbol: LOC101929380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105018.1 | 2 | 187 | |||