siRNA Details
| Gene Symbol: LINC01033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126379.1 | 2 | 266 | |||
| GenBank mRNA | BG181707.1 | - | 4 | 577 | ||
| BG281020.1 | - | 2 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01033| Gene Symbol: LINC01033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126379.1 | 2 | 266 | |||
| GenBank mRNA | BG181707.1 | - | 4 | 577 | ||
| BG281020.1 | - | 2 | 267 | |||