siRNA Details
| Gene Symbol: C5orf66-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105049.1 | 2 | 774 | |||
| NR_105050.1 | 2 | 769 | ||||
| GenBank mRNA | BE785577.1 | - | 2 | 438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf66-AS1| Gene Symbol: C5orf66-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105049.1 | 2 | 774 | |||
| NR_105050.1 | 2 | 769 | ||||
| GenBank mRNA | BE785577.1 | - | 2 | 438 | ||