siRNA Details
| Gene Symbol: LOC389273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134289.1 | 3 | 591 | |||
| NR_134290.1 | 3 | 400 | ||||
| GenBank mRNA | AK130861.1 | - | 3 | 400 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000506021.4 | - | 3 | 400 | Chr. 5 - 10504996 - 10509194 [-] on Build GRCh38 | |

LOC389273
Genomic Map