siRNA Details
| Gene Symbol: CTD-3080P12.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109911.1 | 5 | 1603 | |||
| GenBank mRNA | AK091791.1 | - | 5 | 1603 | ||
| AK097569.1 | - | 1 | 1732 | |||
| BC032469.1 | - | 5 | 1590 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000514376.1 | - | 5 | 1603 | Chr. 5 - 1173141 - 1178605 [-] on Build GRCh38 | |

CTD-3080P12.3
Genomic Map