siRNA Details
| Gene Symbol: LOC101060498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121640.1 | 1 | 446 | |||
| GenBank mRNA | DB230865.1 | - | 1 | 442 | ||
| JN120857.1 | - | 1 | 193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101060498| Gene Symbol: LOC101060498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121640.1 | 1 | 446 | |||
| GenBank mRNA | DB230865.1 | - | 1 | 442 | ||
| JN120857.1 | - | 1 | 193 | |||