siRNA Details
| Gene Symbol: LOC101927100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109875.1 | 2, 3 | 415 | |||
| GenBank mRNA | DA313138.1 | - | 2, 3 | 327 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000508517.1 | - | 2, 3 | 326 | Chr. 5 - 115602244 - 115620278 [+] on Build GRCh38 | |

LOC101927100
Genomic Map