siRNA Details
| Gene Symbol: LOC101927100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109874.1 | 4 | 521 | |||
| NR_109875.1 | 3 | 460 | ||||
| GenBank mRNA | DA313138.1 | - | 3 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927100| Gene Symbol: LOC101927100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109874.1 | 4 | 521 | |||
| NR_109875.1 | 3 | 460 | ||||
| GenBank mRNA | DA313138.1 | - | 3 | 372 | ||