siRNA Details
Gene Symbol: CTD-2194D22.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109912.1 | 4 | 930 | |||
GenBank mRNA | BC039383.1 | - | 4 | 929 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000514569.1 | - | 4 | 929 | Chr. 5 - 1887332 - 1900493 [+] on Build GRCh38 |
