siRNA Details
| Gene Symbol: LOC101927040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110954.1 | 2 | 196 | |||
| GenBank mRNA | AK055332.1 | - | 2 | 197 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000502766.2 | - | 2 | 196 | Chr. 8 - 79769372 - 79871737 [-] on Build GRCh38 | |

LOC101927040
Genomic Map