siRNA Details
| Gene Symbol: LINC01333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105010.1 | 1 | 105 | |||
| GenBank mRNA | BC043537.1 | - | 1 | 105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01333| Gene Symbol: LINC01333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105010.1 | 1 | 105 | |||
| GenBank mRNA | BC043537.1 | - | 1 | 105 | ||