siRNA Details
| Gene Symbol: LOC101927592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104646.1 | 2 | 390 | |||
| GenBank mRNA | BC032874.2 | - | 2 | 390 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000512624.6 | - | 2 | 390 | Chr. 12 - 126915220 - 127060397 [-] on Build GRCh38 | |

LOC101927592
Genomic Map