siRNA Details
Gene Symbol: LINC01016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038989.1 | 3 | 2214 | |||
GenBank mRNA | AK057709.1 | - | 3 | 2212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000506222.6 | - | 3 | 2214 | Chr. 6 - 33889511 - 33896907 [-] on Build GRCh38 |
