siRNA Details
| Gene Symbol: LINC01060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033869.1 | 4 | 308 | |||
| GenBank mRNA | AK095968.1 | - | 5 | 584 | ||
| AY494056.1 | - | 5 | 682 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01060| Gene Symbol: LINC01060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033869.1 | 4 | 308 | |||
| GenBank mRNA | AK095968.1 | - | 5 | 584 | ||
| AY494056.1 | - | 5 | 682 | |||