siRNA Details
| Gene Symbol: LOC101927274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110751.1 | 2 | 1107 | |||
| GenBank mRNA | BC039326.1 | - | 2 | 1111 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000514726.1 | - | 2 | 1110 | Chr. 17 - 51312609 - 51335165 [-] on Build GRCh38 | |

LOC101927274
Genomic Map