siRNA Details
| Gene Symbol: LOC101929412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104606.1 | 1 | 940 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000515243.1 | - | 1 | 940 | Chr. 5 - 10493527 - 10502728 [-] on Build GRCh38 | |

LOC101929412
Genomic Map