siRNA Details
Gene Symbol: LOC101928052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125888.1 | 5 | 852 | |||
GenBank mRNA | BC033991.1 | - | 5 | 857 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000513093.1 | - | 5 | 852 | Chr. 4 - 162022733 - 162047567 [+] on Build GRCh38 |
